missing translation for 'onlineSavingsMsg'
Learn More

Celite∣r 535

Catalog No. AAH377580B
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH377580B 1000 g
AAH377580I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH377580B Supplier Thermo Scientific Chemicals Supplier No. H377580B

CAS: 68855-54-9 | SiO{2}

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Melting Point >450°C
Quantity 1000 g
CAS 68855-54-9
Molecular Formula SiO{2}
Synonym Diatomaceous Earth;Filter aid, flux calcined