missing translation for 'onlineSavingsMsg'
Learn More

Celite(R), 521, Spectrum™ Chemical

Catalog No. 18601523
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 lb.
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601523 500 g Poly Pail
18601526 50 lb. Poly Pail
18601524 2.5 kg Poly Pail
18601525 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601523 Supplier Spectrum Chemical Mfg Cor Supplier No. C1190500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C1190, 91053-39-3

Spectrum™ Chemical Celite, 521 is also known as diatomaceous earth or kieselgur and is a soft sedimentary rock. It is easily crushed into a white powder. It can have an abrasive feel reminiscent of pumice powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 91053-39-3
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Color Pink to Beige
Quantity 500 g
CAS Max % 100%
Packaging Poly Pail
Grade Ungraded
Chemical Name or Material CELITE 521