missing translation for 'onlineSavingsMsg'
Learn More

Celite™ 545 AW Reagent Grade, MilliporeSigma™ Supelco™

Catalog No. 111001610
Change view
Click to view available options
Quantity:
454 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001610 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001610 Supplier Merck Emd Millipore Supplier No. 20199U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00132803
Synonym Diatomaceous earth, flux-calcined; Celite(R) Filter Aid

Specifications

CAS 68855-54-9
Physical Form Powder
Quantity 454 g
Grade Reagent
Product Line Celite

Celite is a registered trademark of Imerys Minerals California, Inc.