missing translation for 'onlineSavingsMsg'
Learn More

Celatom∣r FW-14

Catalog No. p-7030856
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3173330 250 g
AAH317330B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3173330 Supplier Thermo Scientific Chemicals Supplier No. H3173330
Only null left

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 250 g
Chemical Name or Material Celatom™ FW-14
Hazard Category Warning
Hazard Statement GHS H Statement
H351-H373
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P261-P280h
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
EINECSNumber 272-489-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only