missing translation for 'onlineSavingsMsg'
Learn More

Celatom™ Acid-washed for use in Total Dietary Fiber Assay, TDF-100A, MilliporeSigma™ Supelco™

Catalog No. p-7187934
Change view
Click to view available options
Quantity:
50 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012237 50 g
111012236 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012237 Supplier Merck Emd Millipore Supplier No. C865650G
Only null left
Food and Beverages and General Analytical

Specifications

CAS 68855-54-9
Quantity 50 g
MDL Number MFCD00132803
Packaging Poly Bottle
Product Line Celatom

Celatom is a registered trademark of EP Minerals, LLC