missing translation for 'onlineSavingsMsg'
Learn More

Celastrol 98.0+%, TCI America™

Catalog No. C273725MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C273725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C273725MG Supplier TCI America Supplier No. C273725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34157-83-0
Molecular Formula C29H38O4
Molecular Weight (g/mol) 450.62
MDL Number MFCD03424073
InChI Key KQJSQWZMSAGSHN-JJWQIEBTSA-N
Synonym celastrol, tripterine, tripterin, 2r,4as,6as,12br,14as,14br-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid, triterpene, celastrol, celastrus scandens, unii-l8gg98663l, 3-hydroxy-24-nor-2-oxo-1 10 ,3,5,7-friedelatetraen-29-oic acid, 2r,4as,6ar,6as,14as,14br-10-hydroxy-2,4a,6a,6a,9,14a-hexamethyl-11-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid, d:a-friedo-24-noroleana-1 10 ,3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo-, 20alpha
PubChem CID 122724
ChEBI CHEBI:63959
IUPAC Name (2R,4aS,6aS,12bR,14aS,14bR)-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid
SMILES CC1=C(O)C(=O)C=C2C1=CC=C1[C@@]2(C)CC[C@@]2(C)[C@@H]3C[C@@](C)(CC[C@]3(C)CC[C@]12C)C(O)=O

Specifications

Melting Point 203°C
Quantity 25 mg
Formula Weight 450.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Celastrol
RTECSNumber YJ7900000
TSCA No
Recommended Storage Freezer (-20 °C)