missing translation for 'onlineSavingsMsg'
Learn More

Cefuroxime sodium salt , MilliporeSigma™ Supelco™

Catalog No. 111014628
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014628 1 g
111014629 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014628 Supplier Merck Emd Millipore Supplier No. C44171G

Cefuroxime is a semisynthetic cephalosporin, widely used as an antimicrobial agent, in veterinary medication for the treatment of cattle, goats, etc.

Chemical Identifiers

MDL Number MFCD09878727

Specifications

CAS 56238-63-2
Linear Formula C16H15N4 NaO8 S
Quantity 1 g
Formula Weight 446.37
Physical Form Neat