missing translation for 'onlineSavingsMsg'
Learn More

Ceftriaxone sodium hemiheptahydrate

Catalog No. p-7057443
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6600303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6600303 Supplier Thermo Scientific Chemicals Supplier No. J6600303

CAS: 104376-79-6 | C36H46N16Na4O21S6 | 1323.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104376-79-6
Molecular Formula C36H46N16Na4O21S6
Molecular Weight (g/mol) 1323.17
MDL Number MFCD01750405,MFCD01750405,MFCD09749665
InChI Key RGDBKADCOSIOEV-MAODNAKNSA-J
PubChem CID 131843450
IUPAC Name tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Ceftriaxone Sodium Hemiheptahydrate
Physical Form Crystalline Solid
Color White to Yellow
Quantity 1 g
Merck Index 14,1953
Solubility Information Freely soluble in water
Formula Weight 661.6
Chemical Name or Material Ceftriaxone sodium hemiheptahydrate, 850 to 995μg/mg
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 277-930-0
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only