missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceftriaxone Disodium Salt Hemiheptahydrate 98.0+%, TCI America™
Supplier: TCI America C222625G
Specifications
| Ceftriaxone Disodium Salt Hemiheptahydrate | |
| 236°C | |
| C36H46N16Na4O21S6 | |
| 25 g | |
| RGDBKADCOSIOEV-MAODNAKNSA-J | |
| tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate | |
| 131843450 | |
| ≥98.0% (HPLC,T) |
| 104376-79-6 | |
| White-Yellow | |
| MFCD01750405,MFCD01750405,MFCD09749665 | |
| ceftriaxone sodium hydrate | |
| O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 | |
| 1323.17 | |
| 598.53 | |
| Crystalline Powder |
Chemical Identifiers
| 104376-79-6 | |
| 1323.17 | |
| RGDBKADCOSIOEV-MAODNAKNSA-J | |
| 131843450 | |
| O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 |
| C36H46N16Na4O21S6 | |
| MFCD01750405,MFCD01750405,MFCD09749665 | |
| ceftriaxone sodium hydrate | |
| tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate |
Safety and Handling
RTECSNumber : XI0368800
TSCA : No