Learn More
Ceftazidime pentahydrate, 98%, Thermo Scientific Chemicals
$192.69 - $192.69
Chemical Identifiers
| CAS | 78439-06-2 |
|---|---|
| Molecular Formula | C22H22N6O7S2·5H2O |
| Molecular Weight (g/mol) | 636.65 |
| InChI Key | NMVPEQXCMGEDNH-TZVUEUGBSA-N |
| PubChem CID | 131673973 |
| ChEBI | CHEBI:3509 |
| IUPAC Name | 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate |
| SMILES | CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461730050
|
Thermo Scientific Chemicals
461730050 |
5 g |
Each for $192.69
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 78439-06-2 | |
| 636.65 | |
| 131673973 | |
| 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate |
| C22H22N6O7S2·5H2O | |
| NMVPEQXCMGEDNH-TZVUEUGBSA-N | |
| CHEBI:3509 | |
| CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O |
Specifications
| 78439-06-2 | |
| 97.5% min. (HPLC) | |
| 5 g | |
| Solubility in water: slightly soluble. Other solubilities: soluble in alkali,slightly soluble in methanol,dmf,insoluble in acetone,ethanol,chloroform,ether,,ethyl acetate,toluene,dioxane | |
| CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O | |
| 636.65 | |
| CHEBI:3509 | |
| 98% |
| >150.0°C | |
| C22H22N6O7S2·5H2O | |
| 13,1959 | |
| NMVPEQXCMGEDNH-TZVUEUGBSA-N | |
| 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate | |
| 131673973 | |
| 636.65 | |
| Ceftazidime pentahydrate |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is di
GHS Signal Word: Danger
RUO – Research Use Only