missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 98%, Thermo Scientific™

Catalog No. AC461730250
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461730250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461730250 Supplier Thermo Scientific Chemicals Supplier No. 461730250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 78439-06-2
Molecular Formula C22H22N6O7S2·5H2O
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkali,slightly soluble in methanol,dmf,insoluble in acetone,ethanol,chloroform,ether,,ethyl acetate,toluene,dioxane
Chemical Name or Material Ceftazidime pentahydrate

Specifications

Assay Percent Range 97.5% min. (HPLC)
Quantity 25g
Merck Index 13,1959
InChI Key NMVPEQXCMGEDNH-TZVUEUGBSA-N
SMILES CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
IUPAC Name 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
Molecular Weight (g/mol) 636.65
PubChem CID 131673973
ChEBI CHEBI:3509
Formula Weight 636.65
Percent Purity 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is di

RUO – Research Use Only