missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 95%

Catalog No. p-7043239
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6646003 1 g
AAJ6646006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6646003 Supplier Thermo Scientific Chemicals Supplier No. J6646003
Only null left

CAS: 78439-06-2 | C22H32N6O12S2 | 636.648 g/mol

Ceftazidime pentahydrate is an antibacterial agent. It is used especially for Pseudomonas and other gram-negative infections in debilitated patients. It is used in the treatment of patients with infections caused by susceptible strains of organisms in the following diseases: lower respiratory tract infections,skin and skin structure infections, urinary tract infections, bacterial septicemia, bone and joint infections, gynecologic infections, intra abdominal infections (including peritonitis), and central nervous system infections (including meningitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 78439-06-2
Molecular Formula C22H32N6O12S2
Molecular Weight (g/mol) 636.648
MDL Number MFCD00153936
InChI Key NMVPEQXCMGEDNH-TZVUEUGBSA-N
PubChem CID 131673973
ChEBI CHEBI:3509
IUPAC Name 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
SMILES CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O

Specifications

Type Ceftazidime Pentahydrate
Physical Form Powder
Color White
Quantity 1 g
Solubility Information Sparingly soluble in aqueous solution
Formula Weight 636.65
Percent Purity 95%
Chemical Name or Material Ceftazidime pentahydrate
Hazard Category H317-H319-H334
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P363-P501c
RTECSNumber UU2230000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only