missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin Sodium Salt MP Biomedicals

Catalog No. ICN19867783
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19867783 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19867783 Supplier MP Biomedicals Inc Supplier No. 0219867783
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52152-93-9
Molecular Formula C22H21N4NaO8S2+
Molecular Weight (g/mol) 556.54
InChI Key GTZPOHRNWUTXNB-DWBVFMGKSA-O
PubChem CID 124203950
IUPAC Name (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]

Specifications

Quantity 250 mg
Formula Weight 554.5
Chemical Name or Material Cefsulodin Sodium Salt