missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin Sodium Salt Hydrate 95.0+%, TCI America™

Catalog No. p-7125549
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2598200MG 200 mg
C25981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2598200MG Supplier TCI America Supplier No. C2598200MG
Only null left

Chemical Identifiers

CAS 1426397-23-0
Molecular Formula C22H19N4NaO8S2
Molecular Weight (g/mol) 554.52
MDL Number MFCD07793338
InChI Key REACMANCWHKJSM-VKYHOSCXNA-M
PubChem CID 131674802
IUPAC Name sodium 4-carbamoyl-1-{[(6R,7R)-2-carboxylato-8-oxo-7-(2-phenyl-2-sulfonatoacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}pyridin-1-ium
SMILES [Na+].[H][C@]12SCC(C[N+]3=CC=C(C=C3)C(N)=O)=C(N1C(=O)[C@H]2NC(=O)C(C1=CC=CC=C1)S([O-])(=O)=O)C([O-])=O

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 554.52
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cefsulodin Sodium Salt Hydrate
RTECSNumber UU1785000
TSCA Yes
Recommended Storage Refrigerator