missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin sodium salt hydrate, 97%

Catalog No. p-7057458
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66120MD 250 mg
AAJ6612003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66120MD Supplier Thermo Scientific Chemicals Supplier No. J66120MD
Only null left

Cefsulodin sodium (sulcephalosporin, CAS # 52152-93-9) is the salt of the semi-synthetic cephalosporin antibiotic. | CAS: 1426397-23-0 | C22H21N4NaO8S2+ | 556.54 g/mol

Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cefsulodin sodium is a third-generation cephalosporin antibiotic with a narrow range of effectiveness
• Cefsulodin competitively inhibits penicillin binding protein, preventing cell wall synthesis and promoting lysis

Application

• Cefsulodin is mainly effective against Yersinia microorganisms, Pseudomonas aeruginosa, and Staphylococcus aureus
• Cefsulodin is useful for drug resistance studies involving susceptible microbes and for studies related to drug mechanisms
• It is appropriate for selection experiments and other cell culture applications

Chemical Identifiers

CAS 1426397-23-0
MDL Number MFCD07793338
PubChem CID 124203950

Specifications

Type Cefsulodin Sodium Salt
Physical Form Powder
Melting Point 175°C
Color White to Yellow
Quantity 250 mg
Molecular Formula C22H19N4NaO8S2
Merck Index 14,1945
Solubility Information Freely soluble in water
InChI Key REACMANCWHKJSM-VKYHOSCXNA-M
SMILES [Na+].[H][C@]12SCC(C[N+]3=CC=C(C=C3)C(N)=O)=C(N1C(=O)[C@H]2NC(=O)C(C1=CC=CC=C1)S([O-])(=O)=O)C([O-])=O
IUPAC Name sodium 4-carbamoyl-1-{[(6R,7R)-2-carboxylato-8-oxo-7-(2-phenyl-2-sulfonatoacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}pyridin-1-ium
Molecular Weight (g/mol) 554.52
Formula Weight 554.53 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Cefsulodin sodium salt hydrate
Show More Show Less
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 257-692-4
RTECSNumber UU1785000
TSCA Yes
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only