missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin sodium salt, 99% purity, For HPLC analysis, 93%, MP Biomedicals™

Catalog No. icn19867780
Change view
Click to view available options
Quantity:
0.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19867780 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19867780 Supplier MP Biomedicals Inc Supplier No. 0219867780
Only null left

Chemical Identifiers

CAS 52152-93-9
Molecular Formula C22H21N4NaO8S2+
Molecular Weight (g/mol) 556.54
InChI Key GTZPOHRNWUTXNB-DWBVFMGKSA-O
PubChem CID 124203950
IUPAC Name (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]

Specifications

Type Cefsulodin Sodium Salt
Physical Form Crystalline Powder
Color White
Quantity 0.1 g
Assay Percent Range 0.93
Formula Weight 554.5
Percent Purity 99%
Chemical Name or Material Cefsulodin sodium salt, For HPLC analysis