missing translation for 'onlineSavingsMsg'
Learn More

Cefpirome Sulfate 95.0+%, TCI America™

Catalog No. p-7125776
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30071G 1 g
C30075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30071G Supplier TCI America Supplier No. C30071G
Only null left

Chemical Identifiers

CAS 98753-19-6
Molecular Formula C22H24N6O9S3
Molecular Weight (g/mol) 612.647
MDL Number MFCD11864966
InChI Key RKTNPKZEPLCLSF-PXPMWPIZSA-N
Synonym cefpirome sulfate
PubChem CID 134129654
IUPAC Name (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
SMILES CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]

Specifications

Melting Point 200°C
Color White
Quantity 1 g
Formula Weight 612.65
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cefpirome Sulfate
RTECSNumber UW8970000
TSCA No
Recommended Storage Refrigerator