missing translation for 'onlineSavingsMsg'
Learn More

Cefpirome sulfate, 95%, Thermo Scientific Chemicals

A fourth generation cephalosporin antibiotic

$189.77

Chemical Identifiers

CAS 98753-19-6
Molecular Formula C22H24N6O9S3
Molecular Weight (g/mol) 612.647
MDL Number MFCD11864966
InChI Key RKTNPKZEPLCLSF-PXPMWPIZSA-N
PubChem CID 134129654
IUPAC Name (6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
SMILES CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6600103
View Documents Promotion Details
N/A 1 g
Each for $189.77
Only null left
 
Description

Description

A fourth generation cephalosporin antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98753-19-6
612.647
RKTNPKZEPLCLSF-PXPMWPIZSA-N
(6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
C22H24N6O9S3
MFCD11864966
134129654
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]

Specifications

Cefpirome Sulfate
98753-19-6
White
MFCD11864966
RKTNPKZEPLCLSF-PXPMWPIZSA-N
(6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
134129654
95%
Powder
198°C to 202°C
C22H24N6O9S3
Soluble in DMSO
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]
612.647
612.66
Cefpirome sulfate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UW8970000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only