missing translation for 'onlineSavingsMsg'
Learn More

Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals

Catalog No. p-7057549
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6680003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6680003 Supplier Thermo Scientific Chemicals Supplier No. J6680003

A broad spectrum antibiotic

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33564-30-6
Molecular Formula C16H16N3NaO7S2
Molecular Weight (g/mol) 449.428
MDL Number MFCD00079042
InChI Key GNWUOVJNSFPWDD-XMZRARIVSA-M
PubChem CID 23667300
ChEBI CHEBI:3501
IUPAC Name sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]

Specifications

Type Cefoxitin Sodium
Physical Form Powder
Melting Point >160°C
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Boiling Point 843°C
Merck Index 14,1936
Solubility Information Soluble in water or methanol
Formula Weight 449.43
Percent Purity 97+%
Chemical Name or Material Cefoxitin sodium
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 251-574-6
RTECSNumber XI0330500
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only