missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefotetan 98.0+%, TCI America™
$337.40 - $337.40
Chemical Identifiers
| CAS | 69712-56-7 |
|---|---|
| Molecular Formula | C17H17N7O8S4 |
| Molecular Weight (g/mol) | 575.604 |
| MDL Number | MFCD00864983 |
| InChI Key | SRZNHPXWXCNNDU-RHBCBLIFSA-N |
| Synonym | cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid |
| PubChem CID | 53025 |
| IUPAC Name | (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| SMILES | CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2936250MG
|
TCI America
C2936250MG |
250 mg |
Each for $337.40
|
|
|||||
Chemical Identifiers
| 69712-56-7 | |
| 575.604 | |
| SRZNHPXWXCNNDU-RHBCBLIFSA-N | |
| 53025 | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O |
| C17H17N7O8S4 | |
| MFCD00864983 | |
| cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid | |
| (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
Specifications
| 69712-56-7 | |
| C17H17N7O8S4 | |
| 250 mg | |
| SRZNHPXWXCNNDU-RHBCBLIFSA-N | |
| (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 53025 | |
| ≥98.0% (HPLC) | |
| Cefotetan |
| White-Yellow | |
| MFCD00864983 | |
| cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O | |
| 575.604 | |
| 575.60 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XI0330800
TSCA : No
Recommended Storage : Freezer (-20 °C)