missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime sodium salt

Cefotaxime sodium salt, CAS # 64485-93-4, is the sodium salt form of cefotaxime with β-lactamase resistant antibiotic activity. | CAS: 64485-93-4 | C16H16N5NaO7S2 | 477.44 g/mol

Supplier:  Thermo Scientific Chemicals J6269006

Quantity 5 g

 View more versions of this product

Catalog No. AAJ6269006


Only null left

Description

Description

Cefotaxime sodium salt acts as a beta-lactamase resistant antibiotic. It is used as an effective antibacterial against gram-negative bacteria, with the notable exception of pseudomonas and penicillin-resistant strains of streptococcus pneumoniae. It is used to treat infections of the bones, joints, skin, respiratory tract and blood stream.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.

Applications

• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures

Specifications

Chemical Identifiers

64485-93-4
477.44
AZZMGZXNTDTSME-JUZDKLSSSA-M
sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C16H16N5NaO7S2
MFCD00079073
88631411
[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Cefotaxime Sodium Salt
Powder
White to Yellow
C16H16N5NaO7S2
5711411
Soluble in water.
[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
477.44
477.4
Cefotaxime sodium salt
64485-93-4
5 g
MFCD00079073
14,1933
AZZMGZXNTDTSME-JUZDKLSSSA-M
sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
88631411
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P314-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

EINECSNumber : 264-915-9

RTECSNumber : XI0250000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions