Learn More
Thermo Scientific Chemicals Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, CAS # 64485-93-4, is the sodium salt form of cefotaxime with β-lactamase resistant antibiotic activity.
$180.13
Chemical Identifiers
| CAS | 64485-93-4 |
|---|---|
| Molecular Formula | C16H16N5NaO7S2 |
| Molecular Weight (g/mol) | 477.44 |
| MDL Number | MFCD00079073 |
| InChI Key | AZZMGZXNTDTSME-JUZDKLSSSA-M |
| PubChem CID | 88631411 |
| SMILES | [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC454950010
|
Thermo Scientific Chemicals
454950010 |
1 g | Glass Bottle |
Each for $180.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.
Applications
• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures
Chemical Identifiers
| 64485-93-4 | |
| 477.44 | |
| AZZMGZXNTDTSME-JUZDKLSSSA-M | |
| [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 |
| C16H16N5NaO7S2 | |
| MFCD00079073 | |
| 88631411 |
Specifications
| Cefotaxime Sodium Salt | |
| 201.0°C to 210.0°C | |
| C16H16N5NaO7S2 | |
| Solubility in water: soluble. | |
| [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1 | |
| 477.44 | |
| 477.44 | |
| Cefotaxime sodium salt |
| 64485-93-4 | |
| 95% | |
| MFCD00079073 | |
| AZZMGZXNTDTSME-JUZDKLSSSA-M | |
| sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 88631411 | |
| 95% |
RUO – Research Use Only