missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Cefoperazone | |
| 173°C | |
| C25H26N9NaO8S2 | |
| 25 g | |
| [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSC4=NN=NN4C)=C(N3C2=O)C([O-])=O)C2=CC=C(O)C=C2)C(=O)C1=O | |
| 667.65 | |
| CHEBI:3493 | |
| ≥98.0% (HPLC,T) |
| 62893-19-0 | |
| White-Yellow | |
| MFCD00865067 | |
| NCFTXMQPRQZFMZ-WERGMSTESA-M | |
| sodium (6R,7R)-7-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 44187 | |
| 645.67 | |
| Crystalline Powder |
Chemical Identifiers
| 62893-19-0 | |
| 667.65 | |
| NCFTXMQPRQZFMZ-WERGMSTESA-M | |
| CHEBI:3493 | |
| [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSC4=NN=NN4C)=C(N3C2=O)C([O-])=O)C2=CC=C(O)C=C2)C(=O)C1=O |
| C25H26N9NaO8S2 | |
| MFCD00865067 | |
| 44187 | |
| sodium (6R,7R)-7-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Safety and Handling
RTECSNumber : XI0373900
TSCA : No
Recommended Storage : Refrigerator