missing translation for 'onlineSavingsMsg'
Learn More

Cefoperazone sodium salt, MP Biomedicals™

Catalog No. MP219969501
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219969501 1 g
MP219969505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219969501 Supplier MP Biomedicals Inc Supplier No. 0219969501

Cefoperazone sodium salt USP, White powder

  • It is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.
  • Cefoperazone sodium salt is a broad spectrum third generation cephalosporin antibiotic that has similar antimicrobial activity to ceftazidime.
  • It is active against Pseudomonoas aeruginosa and it has enhanced activity against Enterobacteriaceae and Bacteroides sp. in the presence of the beta-lactamase inhibitor sulbactam. Compared to cefotaxime, cefoperazone has greater susceptibility to hydrolysis by some beta-lactamases.

Chemical Identifiers

CAS 62893-20-3
Molecular Formula C25H26N9NaO8S2
Molecular Weight (g/mol) 667.65
MDL Number MFCD07793331
InChI Key NCFTXMQPRQZFMZ-WERGMSTESA-M
PubChem CID 23663974
ChEBI CHEBI:3494

Specifications

Color White
pH 4.5 to 6.5 (1 in 4)
Quantity 1 g
Beilstein 4902135
Synonym Cefobid, Cefoperazone sodium, CPZ, T-1551
Solubility Information Soluble in water (50mg/mL - clear, faint yellow solution).
Grade USP
Physical Form Powder
Assay 870 - 1015 µg/mg
storageNote1 +4°C