missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefditoren pivoxyl

Catalog No. AC463110010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463110010 1 g
AC463110050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463110010 Supplier Thermo Scientific Chemicals Supplier No. 463110010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117467-28-4
Molecular Formula C25H28N6O7S3
Molecular Weight (g/mol) 620.71
MDL Number MFCD00933166
InChI Key AFZFFLVORLEPPO-UHFFFAOYNA-N
IUPAC Name {7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[2-(4-methyl-1,3-thiazol-5-yl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy}methyl 2,2-dimethylpropanoate
SMILES CON=C(C(=O)NC1C2SCC(C=CC3=C(C)N=CS3)=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C1=CSC(N)=N1

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 620.73
Chemical Name or Material Cefditoren pivoxyl
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement If eye irritation persists: Get medical advice/attention.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only