missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefdinir 97.0+%, TCI America™
$195.73 - $490.25
Chemical Identifiers
| CAS | 91832-40-5 |
|---|---|
| Molecular Formula | C14H13N5O5S2 |
| Molecular Weight (g/mol) | 395.408 |
| MDL Number | MFCD00865030 |
| InChI Key | RTXOFQZKPXMALH-GHXIOONMSA-N |
| Synonym | cefdinir |
| PubChem CID | 6915944 |
| ChEBI | CHEBI:3485 |
| IUPAC Name | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| SMILES | C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
Chemical Identifiers
| 91832-40-5 | |
| 395.408 | |
| RTXOFQZKPXMALH-GHXIOONMSA-N | |
| 6915944 | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| C14H13N5O5S2 | |
| MFCD00865030 | |
| cefdinir | |
| CHEBI:3485 | |
| C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
Specifications
| 91832-40-5 | |
| White-Yellow | |
| MFCD00865030 | |
| cefdinir | |
| C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O | |
| 395.408 | |
| CHEBI:3485 | |
| ≥97.0% (HPLC,T) | |
| Cefdinir |
| 190°C | |
| C14H13N5O5S2 | |
| 1 g | |
| RTXOFQZKPXMALH-GHXIOONMSA-N | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 6915944 | |
| 395.41 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XI0367250
TSCA : No