missing translation for 'onlineSavingsMsg'
Learn More

Cefdinir 97.0+%, TCI America™

Catalog No. C31115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31115G 5 g
C31111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31115G Supplier TCI America Supplier No. C31115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91832-40-5
Molecular Formula C14H13N5O5S2
Molecular Weight (g/mol) 395.408
MDL Number MFCD00865030
InChI Key RTXOFQZKPXMALH-GHXIOONMSA-N
Synonym cefdinir
PubChem CID 6915944
ChEBI CHEBI:3485
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 5 g
Formula Weight 395.41
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cefdinir
RTECSNumber XI0367250
TSCA No