missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefdinir 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 190°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 395.41 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cefdinir |
Safety and Handling
| RTECSNumber | XI0367250 |
| TSCA | No |