missing translation for 'onlineSavingsMsg'
Learn More

Cedrol 98.0+%, TCI America™

Catalog No. p-7125684
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28545G 5 g
C285425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28545G Supplier TCI America Supplier No. C28545G

Chemical Identifiers

CAS 77-53-2
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD00062952
InChI Key SVURIXNDRWRAFU-IHZUMSIISA-N
PubChem CID 129317541
SMILES CC1CCC2C13CCC(C(C3)C2(C)C)(C)O

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 273°C
Quantity 5 g
Formula Weight 222.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cedrol
EINECSNumber (4)-0667
RTECSNumber PB7728666
TSCA Yes