missing translation for 'onlineSavingsMsg'
Learn More

Cedrol 98.0+%, TCI America™

Catalog No. c285425g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C285425G 25 g
C28545G 5 g
2 options

Catalog No. C285425G

Supplier: TCI America C285425G

Only null left

Chemical Identifiers

CAS 77-53-2
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD00062952
InChI Key SVURIXNDRWRAFU-IHZUMSIISA-N
PubChem CID 129317541
SMILES CC1CCC2C13CCC(C(C3)C2(C)C)(C)O

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 273°C
Quantity 25 g
Formula Weight 222.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cedrol
EINECSNumber (4)-0667
RTECSNumber PB7728666
TSCA Yes