missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide
For cellular and molecular biology applications
Specifications
| Product Type | Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide |
|---|---|
| Molecular Weight (g/mol) | 1770.29 |
| Content And Storage | -30°C to -10°C |
| Format | Lyophilized Powder |
| Common Name | [CA(1-7)M(2-9)NH2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66741MCR
|
Thermo Scientific™
J66741MCR |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAJ66741EXD
|
Thermo Scientific™
J66741EXD |
0.5 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide | |
| -30°C to -10°C | |
| [CA(1-7)M(2-9)NH2] | |
| HPLC |
| 1770.29 | |
| Lyophilized Powder | |
| RUO - research use only |
RUO â Research Use Only