missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | [CA(1-7)M(2-9)NH2] |
| Content And Storage | -30°C to -10°C |
| Format | Lyophilized Powder |
| For Use With (Application) | Causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes. |
| Molecular Weight (g/mol) | 1770.29 |
| Product Type | Cecropin A (1-7) -Melittin (2-9) Hybrid Peptide |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-NH2 |
RUO – Research Use Only