Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human CEACAM-5 / CD66e Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;CE5,CEACAM-5,CD66e,CEACAM5,CEA,Meconium antigen 100
Catalog No. 501657489
Encompass
Recombinant Protein;Biotinylated Human CEACAM-5 / CD66e Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CEACAM-5, His Tag, Avi Tag (CE5-H82E0) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 75.0 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501657489 Supplier ACROBiosystems Inc Supplier No. CE5H82E025UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Biotinylated Human CEACAM-5 / CD66e Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CEACAM-5, His Tag, Avi Tag (CE5-H82E0) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 75.0 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.