Learn More
Acrobiosystems Recombinant Protein;Human CEACAM-5 / CD66e Protein;HEK293;100UG;CE5,CEACAM-5,CD66e,CEACAM5,CEA,Meconium antigen 100
Supplier: ACROBiosystems Inc CE5H5226100UG
Recombinant Protein;Human CEACAM-5 / CD66e Protein;100UG;Human CEACAM-5 (CE5-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.