missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-glucosamine, 96.0-102.0%, Spectrum™ Chemical

Catalog No. 18611894
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611894 500 g Poly Bottle
18611890 25 g Poly Bottle
18611892 100 g Poly Bottle
18611891 5 kg Poly Bottle
18611893 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611894 Supplier Spectrum Chemical Mfg Cor Supplier No. A1269500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
pH 6.0 to 8.0
Packaging Poly Bottle
Quantity 500 g
Specific Rotation +39.0° to +43.0°
Percent Purity 96 to 102.0%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material N-Acetyl-D-glucosamine

CAUTION: For manufacturing, laboratory or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.