missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional CE TraXero™ Wheel Cleaner Mat

Catalog No. 17549308
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17549308 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17549308 Supplier Vileda Professional Supplier No. 182776

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TraXero accessories expand cleaning performance with a containment mat that stabilizes and protects the floor, plus modular add on bases that extend track length for enhanced wheel cleaning

  • Keeps floor clean from spills
  • Aligns track mechanisms and secures them in place
  • Provides an easy carry option to and from the floor

Specifications

Product Type Wheel Cleaner Mat
Color Gray
For Use With (Equipment) Modular Add On Bases