missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vileda Professional™ CE Multi-Surface Cleaner™
Description
- Designed to clean pipes, hoods and other hard to reach or uneven surfaces
- Manufactured of high performance plastics and stainless steel
- Completely autoclavable and rust free
- Versatile
- Easily bendable to give the tool different shapes to effectively clean any surface
Specifications
Specifications
| Product Type | Multi-Surface Cleaning Tool |
| Color | Blue |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Size | 23 in. |
| Quantity | 1 |
| For Use With (Application) | Pipes, Hoods, Hard to reach Areas |
| Material | Stainless Steel, Plastic |