Learn More
Vileda Professional CE MicroIntensive Duo™ Plus Mop
Single-use mop for IPA rinsing and the removal of disinfectant residue on glass and stainless steel.
$819.10 - $1,031.75
Specifications
| Color Code | White |
|---|---|
| Product Type | Cleanroom Mop |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Color-coded | No |
Description
- High performance dimpled polyester microfiber for effective cleaning results
- High surface coverage with double-sided design
- Effective residue removal on glass and stainless steel
- IPA compatible
- Ideal for damp mopping and pre-preperation cleaning methods
- Streak free cleaning
- Extremely low particle shedding - Up to ISO 4 (GMP A/B)
- Saves time, money, and chemical use
- Choose from laundered (non-sterile) or Gamma Irradiated (sterile) styles
Specifications
| White | |
| Autoclavable | |
| No |
| Cleanroom Mop | |
| Single-use | |
| Microfiber Polyester |