missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vileda Professional CE isoXclean™ MI™ Pad
Description
- Suitable for ISO Class ISO 4-9 / GMP A/B, C
- Application area: isolators, biosafety cabinets, RABS, and other hard to clean areas
- Specialty cleaning of residues on stainless steel and glass surfaces
- High chemical resistance against most disinfectant categories
- Cleanroom laundered and polybagged in CE conditions
- Low friction for ergonomic cleaning and low particle shedding construction
Specifications
Specifications
| Product Type | Isolator Cleaning Tool Cover |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Quantity | 8 Pk. |
| Color-coded | No |
| Material | Polyester, Synthetic Microfiber |