missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional CE isoXclean™ MI™ Pad

Catalog No. 17549304
Change view
Click to view available options
Quantity:
4 Pk.
8 Pk.
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sterility
17549304 4 Pk. Non-sterile
17549305 4 Pk. Sterile
17549306 8 Pk. Non-sterile
17549307 8 Pk. Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17549304 Supplier Vileda Professional Supplier No. 182724

isoXclean pads deliver efficient, low lint cleaning for isolators and controlled environment surfaces.

  • Suitable for ISO Class ISO 4-9 / GMP A/B, C
  • Application area: isolators, biosafety cabinets, RABS, and other hard to clean areas
  • Specialty cleaning of residues on stainless steel and glass surfaces
  • High chemical resistance against most disinfectant categories
  • Cleanroom laundered and polybagged in CE conditions
  • Low friction for ergonomic cleaning and low particle shedding construction

Specifications

Product Type Isolator Cleaning Tool Cover
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 4 Pk.
Color-coded No
Material Polyester, Synthetic Microfiber