missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional CE Duo™ CleanTech™ with Bucket Liner, 6 Pack

Catalog No. 19000684
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19000684 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19000684 Supplier Vileda Professional Supplier No. 172474

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CE Duo™ CleanTech™ Mop is a single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls, and ceilings.

  • Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
  • Adhesive free lamination techniqies are used to prevent additional contamination
  • Excellent microfiber cleaning results have been independently proven
  • Packaged 6 mops per pack, double bagged with bucket liner
  • Reduce packaging waste
  • Reduce amount of time needed to pre-prepare the mops

Specifications

Product Type Cleanroom Mop Head
Color Blue, White
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 6/Pk.
Includes Bucket Liner
For Use With (Application) Floor, wall, and ceiling cleaning and disinfecting
Certifications/Compliance COFC
Color-coded No
Material Laminated Foam
Show More Show Less