Learn More
Vileda Professional™ CE Compact Trolley
Catalog No. 19200840
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19200840 Stainless Steel
1 options
Catalog No. 19200840 Supplier Vileda Professional™ Supplier No. 169843
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Compact, modular, and stowable trolley designed for use with pre-prepared mops and cleaning supplies, that stays inside the space and is adjustable to a variety of uses.
  • Electropolished stainless steel for a contamination free surface
  • Autoclavable
  • Low profile design fits under work stations or tables
  • Fits full sized pre-prep boxes and buckets
  • Reduces the risk of cross contamination by staying inside the controlled space for storage
  • Accessories can be color coded
Specifications
Autoclavable Autoclavable
Finish Electro-polished Stainless Steel
Load Bearing Capacity (English) 150 lb.
Load Bearing Capacity (Metric) 68.04 kg
Material Stainless Steel
No. of Shelves 2