missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDKN1B/CEN12p FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Format | Tube |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 1027 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Quantity | 100 μL |
| Target | Human |
| Form | Liquid |