Learn More
SignalChem Biotech CDK5, Unactive 100ug
Catalog No. 502575577
Encompass
Recombinant full-length human CDK5 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502575577 Supplier SignalChem Biotech Supplier No. C3314G100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human CDK5 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.