missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cdc42 Activation Assay Kit

Catalog No. 17286
Change view
Click to view available options
Quantity:
30 Assays/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17286 30 Assays/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17286 Supplier MilliporeSigma™ Supplier No. 17286

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Rac/cdc42 Assay Reagent (PAK1 PBD, agarose)
  • Mg2+ Lysis/Wash Buffer, 5X
  • 100X GTPγS, 10mM
  • 100X GDP, 100mM
  • Anti-cdc42

Can be used to precipitate cdc42-GTP from cell lysates subjected to various treatments and detection by a cdc42 specific monoclonal antibody

Application: Non-radioactive cdc42 Activation Assay Kit can be used to precipitate cdc42-GTP from cell lysates subjected to various treatments and detection by a cdc42 specific monoclonal antibody

Specifications

Detection Method Chemiluminescence
For Use With (Application) Affinity Binding Assay
Quantity 30 Assays/Kit