missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cdc2-Like Kinase Inhibitor, TG003

$157.40 - $1,013.23

Chemical Identifiers

CAS 300801-52-9
Molecular Formula C13H15NO2S
Synonym Clk Inhibitor; TG003
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64690LB0
View Documents
Thermo Scientific Chemicals
J64690LB0
5 mg N/A
Only null left
 
AAJ64690MF
View Documents
Thermo Scientific Chemicals
J64690MF
50 mg N/A
Only null left
 
Description

Description

Cdc2-Like Kinase Inhibitor, TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members. Reports show that TG003 modifies the regulation of alternative splicing modulated by phosphorylation of SR proteins in vitro, and rescues defects in the development of Xenopus caused by an overabundance of Clk activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
Specifications

Chemical Identifiers

300801-52-9
C13H15NO2S
Clk Inhibitor; TG003

Specifications

5 mg
2°C to 8°C
300801-52-9
249.33g/mol
Light Sensitive
Signal Transduction Reagent
BGVLELSCIHASRV-UHFFFAOYSA-N
1-(3-ethyl-5-methoxy-2,3-dihydro-1,3-benzothiazol-2-ylidene)propan-2-one
Off-white
TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
C13H15NO2S
Clk Inhibitor; TG003
Soluble in DMSO
Solid
CCN1C(SC2=C1C=C(OC)C=C2)=CC(C)=O
249.33
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only