Learn More
ACROBiosystems Inc Recombinant Protein;Mouse CD47 Protein;HEK293;100UG;CD7,CD47,MER6,IAP,OA3
Catalog No. 501658059
Encompass
Recombinant Protein;Mouse CD47 Protein;100UG;Mouse CD47 (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 33-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) at 10 g/mL (100 L/well) can bind Mouse CD47 (Cat. No. CD7-M522b) with a linear range of 0.078-0.625 g/mL (QC tested).
Catalog No. 501658059 Supplier ACROBiosystems Inc Supplier No. CD7M522B100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Mouse CD47 Protein;100UG;Mouse CD47 (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 33-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) at 10 g/mL (100 L/well) can bind Mouse CD47 (Cat. No. CD7-M522b) with a linear range of 0.078-0.625 g/mL (QC tested).