Recombinant Protein;50UG;Human CD47, His Tag, low endotoxin (CD7-H522a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 5 g/mL (100 L/well) can bind Human CD47, His Tag, low endotoxin (Cat. No. CD7-H522a) with a linear range of 2-13 ng/mL (QC tested).;Immobilized Anti-CD47 MAb, Human IgG4 (clone 5F9) at 2 g/mL (100 L/well) can bind Human CD47, His Tag, low endotoxin (Cat. No. CD7-H522a) with a linear range of 0.4-3 ng/mL (Routinely tested).