Learn More
ACROBiosystems Inc ACROBiosystems Cynomolgus / Rhesus macaque CD47 Protein, His Tag
Catalog No. 501957256
Encompass
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 30-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 5 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 4-63 ng/mL (QC tested).;Immobilized Anti-CD47, Human IgG4 at 2 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 0.2-5 ng/mL (Routinely tested).;Anti-CD47 Mab (Human IgG4) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Cynomolgus/Rhesus macaque CD47, H
Catalog No. 501957256 Supplier ACROBiosystems Inc Supplier No. CD7C52H11MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 30-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 5 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 4-63 ng/mL (QC tested).;Immobilized Anti-CD47, Human IgG4 at 2 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 0.2-5 ng/mL (Routinely tested).;Anti-CD47 Mab (Human IgG4) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Cynomolgus/Rhesus macaque CD47, H