missing translation for 'onlineSavingsMsg'
Learn More

Chromium Potassium Sulfate, Dodecahydrate, Technical, Spectrum™ Chemical

Catalog No. 18609774
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609774 100 lb. Amber Glass Bottle
18609771 500 g Amber Glass Bottle
18609772 2.5 kg Amber Glass Bottle
18609773 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609774 Supplier Spectrum Chemical Mfg Cor Supplier No. C1261100LBBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7788-99-0 | 499.4 g/mol

Chemical Identifiers

CAS 7788-99-0

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 100 lb.
Molecular Weight (g/mol) 499.4
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Chromium Potassium Sulfate, Dodecahydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.